missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzylamine, 98%

Product Code. 10394302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10394302 1 g Glass bottle 1g
10405475 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10394302 Supplier Thermo Scientific Supplier No. 365530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-98-4 | C9H14N | 136.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-98-4
Molecular Formula C9H14N
Molecular Weight (g/mol) 136.22
MDL Number MFCD00025575
InChI Key GBSUVYGVEQDZPG-UHFFFAOYSA-O
Synonym 2,4-dimethylbenzylamine, 2,4-dimethylphenyl methanamine, benzenemethanamine, 2,4-dimethyl, benzylamine, 2,4-dimethyl, 2,4-dimethylbenzenemethanamine, omega-amino-psi-cumene, 1-2,4-dimethylphenyl methanamine, omega-amino-1,2,4-trimethylbenzene, rarechem al bw 0350, timtec-bb sbb005795
PubChem CID 66761
IUPAC Name (2,4-dimethylphenyl)methanamine
SMILES CC1=CC=C(C[NH3+])C(C)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2C6H3CH2NH2
Refractive Index 1.5360 to 1.539
Quantity 1 g
Formula Weight 135.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylbenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.