missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzoic acid, 98%

Product Code. p-3778375 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10605602

Brand: Thermo Scientific Acros 160320100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-01-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-01-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002480
InChI Key BKYWPNROPGQIFZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
PubChem CID 11897
ChEBI CHEBI:64811
IUPAC Name 2,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(C)=C1

Specifications

Melting Point 123°C to 127°C
CAS Min % 97.5
Color White
CAS Max % 100.0
Boiling Point 267°C (727.0 mmHg)
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 531
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.