missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzoic acid, 98%

Product Code. 10605602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10605602 10 g Glass bottle 10g
10124030 50 g Glass bottle 50g
10646904 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10605602 Supplier Thermo Scientific Acros Supplier No. 160320100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-01-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-01-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002480
InChI Key BKYWPNROPGQIFZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
PubChem CID 11897
ChEBI CHEBI:64811
IUPAC Name 2,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(C)=C1

Specifications

Melting Point 123°C to 127°C
CAS Min % 97.5
Color White
CAS Max % 100.0
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 531
Boiling Point 267°C (727.0 mmHg)
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.