missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-pentanone, 98%

Product Code. 10458330 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Glass bottle
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 10458330

Brand: Thermo Scientific Acros 159705000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-80-0 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -80°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 124°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH(CH3)2
Refractive Index 1.3990 to 1.402
Quantity 500 mL
Beilstein 01, 703
Specific Gravity 0.8
Solubility Information Solubility in water: insoluble
Viscosity 0.63 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-3-pentanone, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethyl-3-pentanone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.