missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10018199
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10018199 5 g Amber Glass Bottle 5g
10240700 250 mg Amber Glass Bottle 250mg
10610366 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10018199 Supplier Thermo Scientific Maybridge Supplier No. CC65801DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15058-72-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
InChI Key JFXXYKVSLHRBAA-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
PubChem CID 638456
IUPAC Name 2,4-dimethylfuran-3-carboxylic acid
SMILES CC1=COC(=C1C(=O)O)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethyl-3-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.