missing translation for 'onlineSavingsMsg'
Learn More

2,4-dimethyl-3-furaldehyde, Thermo Scientific™

Product Code. 10422560
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422560 1 g Amber Glass Bottle 1g
10595460 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422560 Supplier Thermo Scientific Maybridge Supplier No. CC65804DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 75002-34-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key MGSAYHDYMMHZQE-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furaldehyde, 3-furancarbaldehyde,2,4-dimethyl, 3-furancarboxaldehyde,2,4-dimethyl, 3-furancarboxaldehyde, 2,4-dimethyl-9ci
PubChem CID 21702758
IUPAC Name 2,4-dimethylfuran-3-carbaldehyde
SMILES CC1=COC(=C1C=O)C

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 124.14
Percent Purity 95%
Chemical Name or Material 2, 4-dimethyl-3-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.