missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1-pentene, 99%

Product Code. 11410451
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
This item is not returnable. View return policy

Product Code. 11410451

Brand: Thermo Scientific Alfa Aesar B20002.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2213-32-3 | C7H14 | 98.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2213-32-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00039854
InChI Key LXQPBCHJNIOMQU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1-pentene, 1-pentene, 2,4-dimethyl, acmc-1ch0z, pentene, 2,4-dimethyl, ch3 2chch2c ch3 =ch2, 1-pentene, 2,4-dimethyl-8ci 9ci
PubChem CID 16657
IUPAC Name 2,4-dimethylpent-1-ene
SMILES CC(C)CC(=C)C

Specifications

Density 0.69
Boiling Point 81°C
Flash Point −16°C (−27°F)
Refractive Index 1.3995
Quantity 10 g
UN Number UN3295
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-1-pentene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.