missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-4509884
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10612823 1 g Amber Glass Bottle 1g
11390557 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10612823 Supplier Thermo Scientific Maybridge Supplier No. CC05801DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53137-27-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.187
InChI Key MQGBARXPCXAFRZ-UHFFFAOYSA-N
Synonym 2,4-dimethylthiazole-5-carboxylic acid, dimethyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-5-thiazolecarboxylic acid, 2,4-dimethyl-thiazole-5-carboxylic acid, 2,4-dimethylthiazole-5-carboxylicacid, ksc269g8n, 2,4-dimethylthiazol-5-carboxylic acid, 2,4-dimethylthiazole-5carboxylic acid, 2 4-dimethylthiazole-5-carboxylic acid
PubChem CID 736488
IUPAC Name 2,4-dimethyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 157.19
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethyl-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.