missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 11393246
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393246 5 g 5g
11383246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393246 Supplier Thermo Scientific Alfa Aesar Supplier No. L12584.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33904-03-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00041058
InChI Key CNXSCEPTSXPBTP-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
PubChem CID 2736202
IUPAC Name 1-isothiocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=S)OC

Spécification

Melting Point 51°C to 53°C
Boiling Point 166°C to 167°C (6 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2923
Beilstein 2723226
Sensitivity Moisture sensitive
Formula Weight 195.24
Percent Purity 97%
Chemical Name or Material 2,4-Dimethoxyphenyl isothiocyanate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.