missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10280843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber glass bottle
Glass bottle
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10280843 1 g Glass bottle 1g
10250073 1 g Amber glass bottle 1g
10115280 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10280843 Supplier Thermo Scientific Acros Supplier No. 314290010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84370-87-6
MDL Number MFCD00002005
InChI Key WRAZHLRMDRTLOZ-UHFFFAOYSA-N
PubChem CID 601637
IUPAC Name 1-isocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=O)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 140.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H9NO3
Linear Formula (CH3O)2C6H3NCO
Refractive Index 1.5465 to 1.5485
Quantity 1 g
Synonym 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene
Molecular Weight (g/mol) 179.175
Formula Weight 179.17
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 2, 4-Dimethoxyphenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.