missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoyl chloride, 96%, Thermo Scientific™

Product Code. 11486120
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11486120

Brand: Thermo Scientific Alfa Aesar H27068.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39828-35-8
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017527
InChI Key KVIUXRJCBBXEGJ-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzoylchloride, acmc-20aocn, 2, 4-dimethoxybenzoyl chloride, 2,4-dimethoxy benzoyl chloride, 2,4-dimethoxy-benzoyl chloride, benzoyl chloride,2,4-dimethoxy, 2,4-dimethoxybenzoic acid chloride, 2,4-bis methyloxy benzoyl chloride, 2,4-dimethoxy-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethoxy-9ci
PubChem CID 2734719
IUPAC Name 2,4-dimethoxybenzoyl chloride
SMILES COC1=CC(=C(C=C1)C(=O)Cl)OC

Specifications

Melting Point 57°C to 61°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 200.62
Percent Purity 96%
Chemical Name or Material 2,4-Dimethoxybenzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.