missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzaldehyde, 98%

Product Code. 11434244
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434244 100 g 100g
11424244 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434244 Supplier Thermo Scientific Alfa Aesar Supplier No. A12549.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 613-45-6 | C9H10O3 | 166.176 g/mol

2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

Solubility
Insoluble in water. Soluble in chlorofrom.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-45-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003311
InChI Key LWRSYTXEQUUTKW-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye
PubChem CID 69175
IUPAC Name 2,4-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)C=O)OC

Specifications

Melting Point 67°C to 71°C
Boiling Point 165°C (10 mmHg)
Quantity 100 g
Beilstein 607989
Sensitivity Air Sensitive
Solubility Information Insoluble in water. Soluble in chlorofrom.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.