missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyaniline, 97%, Thermo Scientific™

Product Code. 10606112 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606112 5 g Glass bottle 5g
10261233 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606112 Supplier Thermo Scientific Acros Supplier No. 115350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2735-04-8
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008371
InChI Key GEQNZVKIDIPGCO-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
PubChem CID 17652
IUPAC Name 2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1)N)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color Brown
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11NO2
Linear Formula (CH3O)2C6H3NH2
Quantity 5 g
Beilstein 13, 784
Solubility Information Solubility in water: 7g/L (20°C)
Formula Weight 153.18
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4-Dimethoxyaniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.