missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone, ACROS Organics™

Product Code. 10712053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712053 100 g Glass bottle 100g
10291922 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712053 Supplier Thermo Scientific Acros Supplier No. 115331000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC

Tekniske data

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCH3
Quantity 100 g
Beilstein 08, 267
Formula Weight 180.2
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2', 4'-Dimethoxyacetophenone
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.