missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone, ACROS Organics™

Product Code. 10291922 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10291922

Brand: Thermo Scientific Acros 115330250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCH3
Quantity 25 g
Beilstein 08, 267
Formula Weight 180.2
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2', 4'-Dimethoxyacetophenone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.