missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dihydroxypyridine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxypyridine |
| CAS | 626-03-9 |
| Molecular Formula | C5 H5 N O2 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C5H5NO2/c7-4-1-2-6-5(8)3-4/h1-3H,(H2,6,7,8) |
| SMILES | Oc1ccnc(O)c1 |
| IUPAC Name | pyridine-2,4-diol |
| Molecular Weight (g/mol) | 111.1 |
| Formula Weight | 111.032 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction