missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzophenone, 99%

Product Code. 11464784
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464784 100 g 100g
11474784 500 g 500g
11484784 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464784 Supplier Thermo Scientific Alfa Aesar Supplier No. A13749.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-56-6 | C13H10O3 | 214.22 g/mol

Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-56-6
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002277
InChI Key ZXDDPOHVAMWLBH-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl
PubChem CID 8572
ChEBI CHEBI:34240
IUPAC Name (2,4-dihydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 144°C to 147°C
Density 1.32
Boiling Point 194°C (1 mmHg)
Flash Point 125°C (257°F)
Quantity 100 g
Beilstein 1311566
Merck Index 14,1106
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material 2,4-Dihydroxybenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.