missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic Acid, TRC

Product Code. 30011920
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011920 1 g 1g
30012153 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011920 Supplier Toronto Research Chemicals Supplier No. TRCD6789401G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-86-1
Molecular Formula C7 H6 O4
Molecular Weight (g/mol) 154.12
Synonym 2,4-Dihydroxybenzoic acid, 4-Carboxyresorcinol, 4-Hydroxysalicylic acid, 2,4-DHBA, p-Hydroxysalicylic acid
IUPAC Name 2,4-dihydroxybenzoic acid
SMILES OC(=O)c1ccc(O)cc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H6O4/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,8-9H,(H,10,11)
Formula Weight 154.0266
Chemical Name or Material beta-Resorcylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.