missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic acid, 97%

Product Code. p-112121 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10152810

Brand: Thermo Scientific Acros 114870050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-86-1 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-86-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002451
InChI Key UIAFKZKHHVMJGS-UHFFFAOYSA-N
Synonym beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
PubChem CID 1491
IUPAC Name 2,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)O)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 210.0°C to 215.0°C
Color Beige to Cream or Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula (HO)2C6H3CO2H
Quantity 5 g
Beilstein 10, 377
Merck Index 15, 8278
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol and ether,practically insoluble in benzene
Formula Weight 154.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxybenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.