missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxyacetophenone, 98%

Product Code. 11439962
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11439962

Brand: Thermo Scientific Alfa Aesar A12066.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-84-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-84-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002279
InChI Key SULYEHHGGXARJS-UHFFFAOYSA-N
Synonym 2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
PubChem CID 6990
ChEBI CHEBI:18414
IUPAC Name 1-(2,4-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1O

Specifications

Melting Point 142°C to 146°C
Quantity 50 g
Beilstein 1282505
Merck Index 14,8140
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2',4'-Dihydroxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.