missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxy-6-methylpyrimidine, 97%, Thermo Scientific™

Product Code. 10214670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10214670 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10214670 Supplier Thermo Scientific Acros Supplier No. 114980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 626-48-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 318.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C5H6N2O2
MDL Number MFCD00006028
Quantity 5 g
Beilstein 24, 342
Merck Index 15, 6209
Synonym 6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methyl
Solubility Information Solubility in water: 7g/L water (22°C). Other solubilities: soluble in alkalies,insoluble in ethanol and ether
InChI Key SHVCSCWHWMSGTE-UHFFFAOYSA-N
SMILES CC1=CC(=O)NC(=O)N1
IUPAC Name 6-methyl-1H-pyrimidine-2,4-dione
Molecular Weight (g/mol) 126.12
PubChem CID 12283
ChEBI CHEBI:74733
Formula Weight 126.11
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxy-6-methylpyrimidine, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dihydroxy-6-methylpyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.