missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydro-[1,2,4]triazole-3-thione, TRC

Product Code. 30377805
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377805 500 mg -
30375501 2.5 g 2.5g
30374140 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377805 Supplier Toronto Research Chemicals Supplier No. TRCD679075500MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
SMILES S=C1NC=NN1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2H3N3S/c6-2-3-1-4-5-2/h1H,(H2,3,4,5,6)
Formula Weight 101.0048
Chemical Name or Material 2,4-Dihydro-[1,2,4]triazole-3-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.