missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorotoluene, 98+%

Product Code. 11469143
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469143 5 g 5g
11479143 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469143 Supplier Thermo Scientific Alfa Aesar Supplier No. A11667.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452-76-6 | C7H6F2 | 128.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-76-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.122
MDL Number MFCD00000332
InChI Key MPXDAIBTYWGBSL-UHFFFAOYSA-N
PubChem CID 67983
IUPAC Name 2,4-difluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)F

Specifications

Density 1.121
Boiling Point 114°C to 115°C
Flash Point 15°C (59°F)
Odor Aromatic
Refractive Index 1.449
Quantity 5 g
UN Number UN2388
Beilstein 1931681
Synonym 2,4-difluorotoluene, difluorotoluene, benzene, 2,4-difluoro-1-methyl, 2,4-difluorotolune, 2, 4-difluorotoluene, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluor-1-methylbenzol, pubchem1612, pubchem1613
Formula Weight 128.12
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.