missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorotoluene, 98%, Thermo Scientific™

Product Code. 10368510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10368510 25 g Glass bottle 25g
10472041 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10368510 Supplier Thermo Scientific Acros Supplier No. 232210250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-76-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD00000332
InChI Key MPXDAIBTYWGBSL-UHFFFAOYSA-N
PubChem CID 67983
IUPAC Name 2,4-difluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1500g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3F2
Refractive Index 1.4475 to 1.4495
Quantity 25 g
Specific Gravity 1.15
Synonym 2,4-difluorotoluene, difluorotoluene, benzene, 2,4-difluoro-1-methyl, 2,4-difluorotolune, 2, 4-difluorotoluene, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluor-1-methylbenzol, pubchem1612, pubchem1613
Formula Weight 128.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorotoluene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Difluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.