missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorothiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11446416
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446416 25 g 25g
11436416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446416 Supplier Thermo Scientific Alfa Aesar Supplier No. B24726.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1996-44-7
Molecular Formula C6H4F2S
Molecular Weight (g/mol) 146.155
MDL Number MFCD00203477
InChI Key BICHBFCGCJNCAT-UHFFFAOYSA-N
Synonym 2,4-difluorothiophenol, 2,4-difluorobenzene-1-thiol, 2,4-difluoro thiophenol, benzenethiol, 2,4-difluoro, 2,4-difluorothiophenol technical, 2,4-difluorobenzenet, pubchem2875, acmc-1bpsf, 2,4-difluoro-benzenethiol
PubChem CID 2737073
IUPAC Name 2,4-difluorobenzenethiol
SMILES C1=CC(=C(C=C1F)F)S

Specifications

Density 1.29
Boiling Point 52°C (15 mmHg)
Flash Point 53°C (127°F)
Refractive Index 1.5225
Quantity 25 g
UN Number UN1228
Beilstein 2516728
Sensitivity Air Sensitive
Formula Weight 146.16
Percent Purity 97%
Chemical Name or Material 2,4-Difluorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.