missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetone, 98%

Product Code. 10341113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341113 5 g Glass bottle 5g
10651875 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341113 Supplier Thermo Scientific Acros Supplier No. 376080050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 274682-91-6 | C9H8F2O | 170.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274682-91-6
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD02258866
InChI Key XVHHDTNIOCSTLH-UHFFFAOYSA-N
Synonym 1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
PubChem CID 2734881
IUPAC Name 1-(2,4-difluorophenyl)propan-2-one
SMILES CC(=O)CC1=C(C=C(C=C1)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (F)2C6H3CH2COCH3
Refractive Index 1.4770 to 1.4790
Quantity 5 g
Formula Weight 170.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorophenylacetone
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Difluorophenylacetone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.