missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetic acid, 99%

Product Code. 11484377
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484377 5 g 5g
11494377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484377 Supplier Thermo Scientific Alfa Aesar Supplier No. B20431.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 81228-09-3 | C8H5F2O2 | 171.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81228-09-3
Molecular Formula C8H5F2O2
Molecular Weight (g/mol) 171.12
MDL Number MFCD00009999
InChI Key QPKZIGHNRLZBCL-UHFFFAOYSA-M
Synonym 2,4-difluorophenylacetic acid, 2-2,4-difluorophenyl acetic acid, benzeneacetic acid, 2,4-difluoro, 2,4-difluorophenyl acetic acid, 2,4-ddifluorophenylacetic acid, rarechem al bo 0247, 2,4-difluoro-phenyl-acetic acid, 2,4-difluorobenzeneacetic acid, 2,4-difluorophenylaceticacid
PubChem CID 123581
IUPAC Name 2-(2,4-difluorophenyl)acetic acid
SMILES [O-]C(=O)CC1=CC=C(F)C=C1F

Specifications

Melting Point 117°C to 119°C
Quantity 5 g
Beilstein 3649727
Formula Weight 172.13
Percent Purity 99%
Chemical Name or Material 2,4-Difluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.