missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 10732301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10732301 1 g Glass bottle 1g
10751545 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732301 Supplier Thermo Scientific Acros Supplier No. 303550010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81228-09-3
Molecular Formula C8H5F2O2
Molecular Weight (g/mol) 171.12
MDL Number MFCD00009999
InChI Key QPKZIGHNRLZBCL-UHFFFAOYSA-M
Synonym 2,4-difluorophenylacetic acid, 2-2,4-difluorophenyl acetic acid, benzeneacetic acid, 2,4-difluoro, 2,4-difluorophenyl acetic acid, 2,4-ddifluorophenylacetic acid, rarechem al bo 0247, 2,4-difluoro-phenyl-acetic acid, 2,4-difluorobenzeneacetic acid, 2,4-difluorophenylaceticacid
PubChem CID 123581
IUPAC Name 2-(2,4-difluorophenyl)acetate
SMILES [O-]C(=O)CC1=CC=C(F)C=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 115.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula F2C6H3CH2CO2H
Quantity 1 g
Formula Weight 172.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,4-Difluorophenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.