missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobiphenyl, TRC

Product Code. 30369554
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369554 1 g
30386335 250 mg
30375216 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369554 Supplier Toronto Research Chemicals Supplier No. TRCD4475381G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37847-52-2
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.18
Synonym 2,4-Difluoro-1,1'-biphenyl, 2,4-Difluoro-1-phenylbenzene, 2,4-Difluorobiphenyl
SMILES Fc1ccc(c(F)c1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H8F2/c13-10-6-7-11(12(14)8-10)9-4-2-1-3-5-9/h1-8H
Formula Weight 190.0594
Chemical Name or Material 2,4-Difluorobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.