missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoic-d3 Acid, CDN

Product Code. 30312061
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312061 0.05 g 0.05g
30312190 0.1 g 0.10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312061 Supplier CDN Supplier No. CDND69530.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,5-trideuterio-4,6-difluorobenzoic acid
CAS 1219804-63-3
Molecular Formula C7 2H3 H F2 O2
Quantity 0.05 g
InChI Formula InChI=1 S/C7H4F2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3 H,(H,10,11)/i1D,2 D,3 D
SMILES [2 H]c1c([2 H])c(C(=O)O)c(F)c([2 H])c1F
IUPAC Name 2,3,5-trideuterio-4,6-difluorobenzoic acid
Molecular Weight (g/mol) 161.121
Formula Weight 161.037 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.