missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Difluorobenzhydrol, TRC

Product Code. 30387749
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387749 1 g -
30376803 25 g -
30385095 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387749 Supplier Toronto Research Chemicals Supplier No. TRCD4454451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153877-53-3
Molecular Formula C13 H10 F2 O
Molecular Weight (g/mol) 220.215
Synonym 2-Fluoro-alpha-(4-fluorophenyl)-benzenemethanol, (2-Fluorophenyl)(4-fluorophenyl)methanol
IUPAC Name (2-fluorophenyl)-(4-fluorophenyl)methanol
SMILES OC(c1ccc(F)cc1)c2ccccc2F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H10F2O/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8,13,16H
Formula Weight 220.07
Chemical Name or Material 2,4'-Difluorobenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.