missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzenesulfonamide, 96%, Thermo Scientific Chemicals

Product Code. 15491668
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491668 5 g 5g
15401678 250 mg 250mg
15481668 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491668 Supplier Thermo Scientific Alfa Aesar Supplier No. H55437.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13656-60-5 | Molecular Formula: C6H5F2NO2S | Molecular Weight: 193.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13656-60-5
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.168
MDL Number MFCD01320743
InChI Key PTHLPYKPCQLAOF-UHFFFAOYSA-N
PubChem CID 574964
IUPAC Name 2,4-difluorobenzenesulfonamide
SMILES C1=CC(=C(C=C1F)F)S(=O)(=O)N

Specifications

Melting Point 154°C to 158°C
Quantity 5 g
Synonym 2,4-difluorobenzene-1-sulfonamide, 2,4-difluorobenzenesulphonamide, benzenesulfonamide, 2,4-difluoro, benzenesulfonamide,2,4-difluoro, buttpark 23\07-80, 2,4-difluoro-1-benzenesulfonamide, 2,4-difluorobenzenesulfoneamide, pubchem11764, enamine_001177, acmc-1c82s
Formula Weight 193.17
Percent Purity 96%
Chemical Name or Material 2,4-Difluorobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.