missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzeneboronic acid, 97%

Product Code. 11460288
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460288 25 g 25g
11440288 1 g 1g
11450288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460288 Supplier Thermo Scientific Alfa Aesar Supplier No. B23821.14
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 144025-03-6 | C6H5BF2O2 | 157.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144025-03-6
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.91
MDL Number MFCD01318998
InChI Key QQLRSCZSKQTFGY-UHFFFAOYSA-N
Synonym 2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
PubChem CID 2734334
IUPAC Name (2,4-difluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1F

Specifications

Melting Point 247°C to 250°C
Quantity 25 g
Beilstein 8616257
Formula Weight 157.91
Percent Purity 97%
Chemical Name or Material 2,4-Difluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.