missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzeneacetic Acid, TRC

Product Code. 30374313
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374313 25 g
30382994 5 g
30383849 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374313 Supplier Toronto Research Chemicals Supplier No. TRCD44540525G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81228-09-3
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
Synonym 2,4-Difluorophenylacetic Acid, 2-(2,4-Difluorophenyl)acetic Acid
IUPAC Name 2-(2,4-difluorophenyl)acetic acid
SMILES OC(=O)Cc1ccc(F)cc1F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H6F2O2/c9-6-2-1-5(3-8(11)12)7(10)4-6/h1-2,4H,3H2,(H,11,12)
Formula Weight 172.0336
Chemical Name or Material 2,4-Difluorobenzeneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.