missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzaldehyde, 98%

Product Code. 10097342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10097342 25 g Glass bottle 25g
10757393 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10097342 Supplier Thermo Scientific Acros Supplier No. 271470250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1550-35-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010326
InChI Key WCGPCBACLBHDCI-UHFFFAOYSA-N
PubChem CID 73770
IUPAC Name 2,4-difluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 2.0°C to 3.0°C
Color Colorless
Density 1.2900g/mL
Boiling Point 65.0°C to 66.0°C (17.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CHO
Refractive Index 1.4975 to 1.4995 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.29
Synonym 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde
Formula Weight 142.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.