missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoroacetophenone, 99%

Product Code. 10519263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10519263 50 g 50g
10195190 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10519263 Supplier Thermo Scientific Acros Supplier No. 246770500
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 364-83-0 | C8H6F2O | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-83-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.12
MDL Number MFCD00151261
InChI Key QEWHNJPLPZOEKU-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
PubChem CID 67770
IUPAC Name 1-(2,4-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 80.0°C to 81.0°C (25.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COCH3
Refractive Index 1.4870 to 1.4890
Quantity 50 g
Specific Gravity 1.23
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble/miscible in polar solvents,insoluble in non-polar solvents
Formula Weight 156.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2', 4'-Difluoroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2',4'-Difluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.