missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoroacetophenone, 98%

Product Code. 11475453
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475453 25 g 25g
11465453 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475453 Supplier Thermo Scientific Alfa Aesar Supplier No. A13420.14
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 364-83-0 | C8H6F2O | 156.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-83-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00151261
InChI Key QEWHNJPLPZOEKU-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
PubChem CID 67770
IUPAC Name 1-(2,4-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)F

Specifications

Density 1.232
Boiling Point 80°C to 81°C (25 mmHg)
Flash Point 66°C (150°F)
Odor Characteristic
Refractive Index 1.488
Quantity 25 g
Beilstein 1940741
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 2',4'-Difluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.