missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-nitrophenol, 98%

Product Code. 15454757
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454757 25 g 25g
15444757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454757 Supplier Thermo Scientific Alfa Aesar Supplier No. H32081.14
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 113512-57-5 | C6H3F2NO3 | 175.091 g/mol

2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

Solubility
Sparingly soluble in water but readily soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113512-57-5
Molecular Formula C6H3F2NO3
Molecular Weight (g/mol) 175.091
MDL Number MFCD08443942
InChI Key SMRYCTJAGPDVEH-UHFFFAOYSA-N
Synonym 2,4-difluoro-5-nitrobenzenol, phenol, 2,4-difluoro-5-nitro, difluoronitrobenzenol, acmc-209wyr, 2,4-difluoro-5-hydroxynitrobenzene
PubChem CID 10921024
IUPAC Name 2,4-difluoro-5-nitrophenol
SMILES C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-]

Specifications

Melting Point 112°C to 120°C
Quantity 25 g
Solubility Information Sparingly soluble in water but readily soluble in organic solvents.
Formula Weight 175.09
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-5-nitrophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.