missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-(2-oxoimidazolidin-1-yl)benzoic Acid, TRC

Product Code. 30381073
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381073 100 mg -
30381691 50 mg 50mg
30375476 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381073 Supplier Toronto Research Chemicals Supplier No. TRCD450083100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1178351-23-9
Molecular Formula C10H8N2O3F2
Molecular Weight (g/mol) 242.17
IUPAC Name 2,4-difluoro-5-(2-oxoimidazolidin-1-yl)benzoic acid
SMILES OC(=O)c1cc(N2CCNC2=O)c(F)cc1F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8F2N2O3/c11-6-4-7(12)8(3-5(6)9(15)16)14-2-1-13-10(14)17/h3-4H,1-2H2,(H,13,17)(H,15,16)
Formula Weight 242.0503
Chemical Name or Material 2,4-Difluoro-5-(2-oxoimidazolidin-1-yl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.