missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzoic acid, 98+%

Product Code. 11456430
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456430 5 g 5g
11401090 1 g 1g
11466430 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456430 Supplier Thermo Scientific Alfa Aesar Supplier No. B25597.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 178974-97-5 | C8H6F2O3 | 188.13 g/mol

2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178974-97-5
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD02258905
InChI Key KWLIGHXPUYUTBH-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxy-benzoic acid, 2,4-difluoro-3-methoxylbenzoic acid, 2,4-difluoro-3-methoxy benzoic acid, pubchem4941, acmc-1cabj, 3-methoxy-2,4-difluorobenzoic acid, benzoic acid,2,4-difluoro-3-methoxy
PubChem CID 3780555
IUPAC Name 2,4-difluoro-3-methoxybenzoic acid
SMILES COC1=C(F)C=CC(C(O)=O)=C1F

Specifications

Melting Point 193°C to 195°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 188.13
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluoro-3-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.