missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-1-nitrobenzene, 98%

Product Code. 11480783
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480783 25 g 25g
11490783 100 g 100g
11400793 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480783 Supplier Thermo Scientific Alfa Aesar Supplier No. A13249.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 446-35-5 | C6H3F2NO2 | 159.09 g/mol

2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-35-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007050
InChI Key RJXOVESYJFXCGI-UHFFFAOYSA-N
Synonym 2,4-difluoronitrobenzene, benzene, 2,4-difluoro-1-nitro, 1-nitro-2,4-difluorobenzene, 1,3-difluoro-4-nitrobenzene, unii-6398y1d7js, 2,4-difluoro-1-nitro-benzene, pubchem2288, 2,4-difluornitrobenzol, 2,4-difluoronitrobenzol, acmc-1akft
PubChem CID 67967
IUPAC Name 2,4-difluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C=C1F

Specifications

Melting Point 9°C to 10°C
Density 1.45
Boiling Point 203°C to 204°C
Flash Point 90°C (194°F)
Refractive Index 1.513
Quantity 25 g
UN Number UN2810
Beilstein 2048034
Solubility Information Insoluble in water.
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-1-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.