missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorothiophenol, 97%

Product Code. 11459533
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459533 1 g 1g
11469533 5 g 5g
11479533 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459533 Supplier Thermo Scientific Alfa Aesar Supplier No. A12384.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-41-4 | C6H4Cl2S | 179.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-41-4
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041421
InChI Key FGBVJFREPSJSNG-UHFFFAOYSA-N
Synonym 2,4-dichlorothiophenol, 2,4-dichlorobenzene-1-thiol, benzenethiol, 2,4-dichloro, 2,4-dichloro thiophenol, pubchem6798, 2,4 dichlorothiophenol, acmc-1buvb, 2,4-dichlorothio-phenol, 2,4-dichloro-benzenethiol, #
PubChem CID 70730
IUPAC Name 2,4-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)Cl)S

Specifications

Melting Point ∼15°C
Density 1.44
Boiling Point 60°C to 62°C (1.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.619
Quantity 1 g
Beilstein 2042868
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.