missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroquinazoline, 97%

Product Code. 15537529 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15537529 5 g 5g
15547529 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15537529 Supplier Thermo Scientific Acros Supplier No. 460730050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 607-68-1 | C8H4Cl2N2 | 199.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-68-1
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.03
MDL Number MFCD00091950
InChI Key TUQSVSYUEBNNKQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-quinazoline, quinazoline, 2,4-dichloro, dichloroquinazoline, 2,4-dichlorchinazolin, pubchem9668, 2,4-dichloroquinazol, 2,4dichloroquinazoline, acmc-209mlw, 2,4-dichloroquinazoline;, 2, 4-dichloro-quinazoline
PubChem CID 252886
IUPAC Name 2,4-dichloroquinazoline
SMILES ClC1=NC(Cl)=C2C=CC=CC2=N1

Specifications

Melting Point 115.0°C to 121.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 199.04
Percent Purity 98%
Chemical Name or Material 2,4-Dichloroquinazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.