missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrimidine, 98+%

Product Code. 11412108
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11412108

Brand: Thermo Scientific Alfa Aesar A15131.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3934-20-1 | C4H2Cl2N2 | 148.974 g/mol

2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3934-20-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006061
InChI Key BTTNYQZNBZNDOR-UHFFFAOYSA-N
PubChem CID 77531
IUPAC Name 2,4-dichloropyrimidine
SMILES C1=CN=C(N=C1Cl)Cl

Specifications

Melting Point 57°C to 62°C
Boiling Point 101°C (23 mmHg)
Quantity 250 g
Beilstein 110911
Sensitivity Moisture sensitive
Synonym pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
Solubility Information Soluble in water (partly),methanol,chloroform,and ethyl acetate.
Formula Weight 148.98
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichloropyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.