missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylhydrazine hydrochloride, 99%, Thermo Scientific™

Product Code. 10592951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10592951 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10592951 Supplier Thermo Scientific Acros Supplier No. 166740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5446-18-4
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C6H7Cl3N2
Linear Formula Cl2C6H3NHNH2·HCl
MDL Number MFCD00012929
Quantity 25g
Beilstein 15, I, 107
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 213.486
PubChem CID 2723905
Formula Weight 213.49
Percent Purity 99%
Chemical Name or Material 2, 4-Dichlorophenylhydrazine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorophenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.