missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylhydrazine hydrochloride, 98%, Thermo Scientific™

Product Code. 11404363
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404363 10 g 10g
11414363 50 g 50g
11424363 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404363 Supplier Thermo Scientific Alfa Aesar Supplier No. A10943.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5446-18-4
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012929
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
PubChem CID 2723905
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl

Specifications

Melting Point ∼212°C (decomposition)
Quantity 10 g
Beilstein 3708828
Formula Weight 213.49
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorophenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.