missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylacetic acid, 98+%

Product Code. 11439373
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439373 5 g 5g
11449373 25 g 25g
11459373 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439373 Supplier Thermo Scientific Alfa Aesar Supplier No. A11791.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19719-28-9 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19719-28-9
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004318
InChI Key GXMWLJKTGBZMBH-UHFFFAOYSA-N
PubChem CID 88209
IUPAC Name 2-(2,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)CC(=O)O

Specifications

Melting Point 128°C to 130°C
Odor Odorless
Quantity 5 g
Beilstein 1948767
Synonym 2,4-dichlorophenylacetic acid, 2-2,4-dichlorophenyl acetic acid, benzeneacetic acid, 2,4-dichloro, 2,4-dichloro-phenyl-acetic acid, 2,4-dichlorophenyl acetic acid, 2 ,4-dichlorophenylacetic acid, zlchem 477, pubchem15026, acmc-209f0x
Formula Weight 205.04
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichlorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.