missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isothiocyanate, 97%

Product Code. 11479606
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479606 5 g 5g
11489606 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479606 Supplier Thermo Scientific Alfa Aesar Supplier No. B20178.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6590-96-1 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-96-1
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00037827
InChI Key WVBNZZHGECFCSH-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
PubChem CID 81066
IUPAC Name 2,4-dichloro-1-isothiocyanatobenzene
SMILES ClC1=CC=C(N=C=S)C(Cl)=C1

Specifications

Melting Point 36°C to 39°C
Boiling Point 130°C to 133°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 777072
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.