missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isocyanate, 96%

Product Code. 11327546
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327546 25 g 25g
11307546 1 g 1g
11317546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327546 Supplier Thermo Scientific Alfa Aesar Supplier No. L11089.14
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2612-57-9 | C7H3Cl2NO | 188.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2612-57-9
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002001
InChI Key OLBJNSPBWLCTOT-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl isocyanate, 2,4-dichlorophenylisocyanate, benzene, 2,4-dichloro-1-isocyanato, 2,4-dichlorophenyl isocyantae, 2,4-dichlorobenzenisocyanate, acmc-1cg9n, 2,4-dichlorphenyl isocyanate, 2,4-dichloro phenyl isocyanate, #
PubChem CID 75789
IUPAC Name 2,4-dichloro-1-isocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)N=C=O

Specifications

Melting Point 57°C to 61°C
Boiling Point 80°C (1 mmHg)
Odor Irritating
Quantity 25 g
UN Number UN2250
Beilstein 2090780
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 96%
Chemical Name or Material 2,4-Dichlorophenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.