missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenoxyacetic acid, 97%

Product Code. 10730975 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10730975

Brand: Thermo Scientific Acros 113635000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-75-7
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
MDL Number MFCD00004300
InChI Key OVSKIKFHRZPJSS-UHFFFAOYSA-N
Synonym 2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
PubChem CID 1486
ChEBI CHEBI:28854
IUPAC Name 2-(2,4-dichlorophenoxy)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 137°C to 141°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula Cl2C6H3OCH2CO2H
Quantity 500 g
Formula Weight 221.04
Percent Purity ≥97.0%
Physical Form Crystalline Flakes, Crystalline Powder or Powder
Chemical Name or Material 2, 4-Dichlorophenoxyacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorophenoxyacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Dichlorophenoxyacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.